Learn More
Waters Corp Waters Corporation Guard Cartridge, Shield RP18, 19 × 10 mm, 10 μm
Catalog No. 50780559 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XTerra Shield RP18 Prep Guard Cartridge is designed to protect analytical columns, ensuring high-efficiency separations and extended column lifetime. It features first-generation hybrid particle stationary phases, combining the efficiency of silica with the stability of polymers. This cartridge is particularly suited for applications requiring a wide pH range and offers excellent water wettability even in 100% aqueous mobile phases.
  • Combines efficiency of silica with polymer stability.
  • Designed for high-efficiency separations.
  • Offers a wide pH range for versatile use.
  • Provides high mechanical strength and excellent peak shape for bases.
  • Features an embedded polar group for optimal selectivity and water wettability.
  • Facilitates easy scale-up from analytical to preparative chromatography.
Catalog No. 50-780-559 Supplier Waters Corp Supplier No. 186001036
The XTerra Shield RP18 Prep Guard Cartridge is designed to protect analytical columns, ensuring high-efficiency separations and extended column lifetime. It features first-generation hybrid particle stationary phases, combining the efficiency of silica with the stability of polymers. This cartridge is particularly suited for applications requiring a wide pH range and offers excellent water wettability even in 100% aqueous mobile phases.
  • Combines efficiency of silica with polymer stability.
  • Designed for high-efficiency separations.
  • Offers a wide pH range for versatile use.
  • Provides high mechanical strength and excellent peak shape for bases.
  • Features an embedded polar group for optimal selectivity and water wettability.
  • Facilitates easy scale-up from analytical to preparative chromatography.