Learn More
Waters Corp XTerra Shield RP18 Prep Guard Cartridge, 125Å, 10 µm, 10 mm X 10 mm, 1/pk
Catalog No. 50780531 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Preparative RP18 columns are hybrid-based, reversed-phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column. This product needs to be used in conjunction with other parts. Please see the section below on related parts for more information
Catalog No. 50-780-531 Supplier Waters Corp Supplier No. 186001007
XTerra Preparative RP18 columns are hybrid-based, reversed-phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column. This product needs to be used in conjunction with other parts. Please see the section below on related parts for more information