Learn More
Waters Corp Waters Prep Column, Shield RP18, 10 μm, 7.8 mm, 150 mm
Catalog No. 50780594 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Preparative RP18 columns are hybrid-based, reversed-phase C18 columns that incorporate embedded polar group technology. These first-generation columns improve peak shape performance for highly basic compounds and provide superior pH stability compared to silica-based columns.
  • Hybrid-based, reversed-phase C18 column
  • Incorporates embedded polar group technology
  • Improves peak shape performance for highly basic compounds
  • Provides superior pH stability (pH range 2–12)
  • High efficiency and mechanical strength
  • Excellent water wettability, even in 100% aqueous mobile phases
Catalog No. 50-780-594 Supplier Waters Corp Supplier No. 186001162
Add to Cart
Add to Cart
XTerra Preparative RP18 columns are hybrid-based, reversed-phase C18 columns that incorporate embedded polar group technology. These first-generation columns improve peak shape performance for highly basic compounds and provide superior pH stability compared to silica-based columns.
  • Hybrid-based, reversed-phase C18 column
  • Incorporates embedded polar group technology
  • Improves peak shape performance for highly basic compounds
  • Provides superior pH stability (pH range 2–12)
  • High efficiency and mechanical strength
  • Excellent water wettability, even in 100% aqueous mobile phases