Learn More
Waters Corp Waters XTerra OBD Prep Column, Shield RP18, 125Å, 10 μm, 19 mm X 150 mm
Catalog No. 50781033 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XTerra Preparative RP18 column is a hybrid-based, reversed-phase C18 column with patented embedded polar group technology, ensuring superior pH stability and excellent peak shape for basic compounds. It combines silica and polymer-bonded phases via Hybrid Particle Technology (HPT) for high mechanical strength and efficient scale-up. Manufactured under strict cGMP and ISO 9002 conditions, it guarantees high quality and consistent analytical results across a wide pH and temperature range.
  • Hybrid-based, reversed-phase C18 chemistry.
  • Embedded polar group technology optimizes peak shape.
  • Superior pH stability.
  • Hybrid Particle Technology (HPT) for robust performance.
  • Stable across low- and high-pH purifications.
  • Provides excellent peak shape for bases.
  • cGMP and ISO 9002 manufactured.
  • Operates from pH 2 to 12 and up to 60°C.
Catalog No. 50-781-033 Supplier Waters Corp Supplier No. 186002257
The XTerra Preparative RP18 column is a hybrid-based, reversed-phase C18 column with patented embedded polar group technology, ensuring superior pH stability and excellent peak shape for basic compounds. It combines silica and polymer-bonded phases via Hybrid Particle Technology (HPT) for high mechanical strength and efficient scale-up. Manufactured under strict cGMP and ISO 9002 conditions, it guarantees high quality and consistent analytical results across a wide pH and temperature range.
  • Hybrid-based, reversed-phase C18 chemistry.
  • Embedded polar group technology optimizes peak shape.
  • Superior pH stability.
  • Hybrid Particle Technology (HPT) for robust performance.
  • Stable across low- and high-pH purifications.
  • Provides excellent peak shape for bases.
  • cGMP and ISO 9002 manufactured.
  • Operates from pH 2 to 12 and up to 60°C.