Learn More
Waters Corp XTerra Shield RP18 Prep Column, 125Å, 5 µm, 7.8 mm X 150 mm, 1/pk
Catalog No. 50780706 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Preparative RP18 columns are hybrid-based, reversed-phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.
Catalog No. 50-780-706 Supplier Waters Corp Supplier No. 186001477
Add to Cart
Add to Cart
XTerra Preparative RP18 columns are hybrid-based, reversed-phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.