Learn More
Waters Corp Waters HPLC Column, Shield RP18, 5 μm, 50 x 100 mm
Catalog No. 50781025 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra HPLC Columns, featuring Hybrid Particle Technology (HPT), merge silica and polymer benefits for enhanced speed, resolution, and pH 1–12 stability. These robust, efficient columns support analytical and preparative chromatography with direct scale–up, particularly for basic compounds.
  • Wide pH range (1–12) for robust separations.
  • HPT delivers high efficiency and sharp peaks for basic compounds.
  • High strength and chemical stability extend column lifetime, especially at high pH (>8.0).
  • Shield technology improves basic analyte peak shape.
  • OBD design enables direct scale–up.
  • Versatile C18, C8, and Phenyl chemistries.
  • MS compatibility: sharp peaks, high sensitivity.
  • eCord/eConnect tags provide traceability and usage data.
Catalog No. 50-781-025 Supplier Waters Corp Supplier No. 186002224
Add to Cart
Add to Cart
Request a Quote
XTerra HPLC Columns, featuring Hybrid Particle Technology (HPT), merge silica and polymer benefits for enhanced speed, resolution, and pH 1–12 stability. These robust, efficient columns support analytical and preparative chromatography with direct scale–up, particularly for basic compounds.
  • Wide pH range (1–12) for robust separations.
  • HPT delivers high efficiency and sharp peaks for basic compounds.
  • High strength and chemical stability extend column lifetime, especially at high pH (>8.0).
  • Shield technology improves basic analyte peak shape.
  • OBD design enables direct scale–up.
  • Versatile C18, C8, and Phenyl chemistries.
  • MS compatibility: sharp peaks, high sensitivity.
  • eCord/eConnect tags provide traceability and usage data.