Learn More
Waters Corp XTerra Shield RP18 OBD Prep Column, 125Å, 5 µm, 50 mm X 50 mm, 1/pk
Supplier:  Waters Corp 186002220
Encompass_Preferred
XTerra Preparative RP18 columns are hybrid-based, reversed-phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.
Catalog No. 50-781-021
Add to Cart
Request a Quote