Learn More
Waters Corp Waters Preparative Column, Shield RP18, 5 μm, 30 mm × 75 mm, 125 Å
Catalog No. 50781097 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Preparative RP18 columns are hybrid-based, reversed-phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.
  • Hybrid-based, reversed-phase C18 columns.
  • Incorporates embedded polar group technology to improve peak shape performance for highly basic compounds.
  • Provides superior pH stability compared to a silica-based column.
Catalog No. 50-781-097 Supplier Waters Corp Supplier No. 186002389
Add to Cart
Add to Cart
Request a Quote
XTerra Preparative RP18 columns are hybrid-based, reversed-phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.
  • Hybrid-based, reversed-phase C18 columns.
  • Incorporates embedded polar group technology to improve peak shape performance for highly basic compounds.
  • Provides superior pH stability compared to a silica-based column.