Learn More
Waters Corp Waters Guard Cartridge, Shield RP8 Prep, 10 μm, 19 x 10 mm
Catalog No. 50780560 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge features a hybrid particle stationary phase, combining high efficiency of silica with expanded pH range of a polymer. It is designed for high mechanical strength, superior peak shape, and a wide pH operating range, making it a rugged and versatile choice for chromatography.
  • Hybrid particle (silica co-polymer) stationary phase
  • High efficiency separations with expanded pH range (1–12)
  • High mechanical strength and excellent peak shape for bases
  • Embedded polar group for optimal peak shape and unique selectivity
  • Excellent water wettability
  • Compatible with UV, fluorescence, electrochemical, and RI detectors
  • Easy scale-up from analytical to preparative chromatography
  • Increased mass loading (up to 60x) at high pH
  • 10 μm particle size
  • 19 x 10 mm dimensions
Catalog No. 50-780-560 Supplier Waters Corp Supplier No. 186001037
This guard cartridge features a hybrid particle stationary phase, combining high efficiency of silica with expanded pH range of a polymer. It is designed for high mechanical strength, superior peak shape, and a wide pH operating range, making it a rugged and versatile choice for chromatography.
  • Hybrid particle (silica co-polymer) stationary phase
  • High efficiency separations with expanded pH range (1–12)
  • High mechanical strength and excellent peak shape for bases
  • Embedded polar group for optimal peak shape and unique selectivity
  • Excellent water wettability
  • Compatible with UV, fluorescence, electrochemical, and RI detectors
  • Easy scale-up from analytical to preparative chromatography
  • Increased mass loading (up to 60x) at high pH
  • 10 μm particle size
  • 19 x 10 mm dimensions