Learn More
Waters Corp XTerra Shield RP8 Prep Guard Cartridge, 125Å, 5 µm, 10 mm X 10 mm, 1/pk
Catalog No. 50780532 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Preparative RP8 columns are hybrid-based, reversed-phase C8 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column. This product needs to be used in conjunction with other parts. Please see the section below on related parts for more information.
Catalog No. 50-780-532 Supplier Waters Corp Supplier No. 186001008
XTerra Preparative RP8 columns are hybrid-based, reversed-phase C8 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column. This product needs to be used in conjunction with other parts. Please see the section below on related parts for more information.