Learn More
Waters Corp Waters XTerra Guard Cartridge, Shield RP8, 5 micron, 7.8 x 10 mm
Catalog No. 50780603 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XTerra Guard Cartridge is designed for high-performance liquid chromatography applications. This guard cartridge features a Shield RP8 type, a 5 μm particle size, 125 Å pore size, and dimensions of 7.8 x 10 mm.
  • Designed for high-performance liquid chromatography applications.
  • Features Shield RP8 type.
  • Has a 5 μm particle size.
  • Includes a 125 Å pore size.
  • Measures 7.8 x 10 mm.
  • Requires a 7.8 x 10 mm cartridge holder.
Catalog No. 50-780-603 Supplier Waters Corp Supplier No. 186001171
The XTerra Guard Cartridge is designed for high-performance liquid chromatography applications. This guard cartridge features a Shield RP8 type, a 5 μm particle size, 125 Å pore size, and dimensions of 7.8 x 10 mm.
  • Designed for high-performance liquid chromatography applications.
  • Features Shield RP8 type.
  • Has a 5 μm particle size.
  • Includes a 125 Å pore size.
  • Measures 7.8 x 10 mm.
  • Requires a 7.8 x 10 mm cartridge holder.