Learn More
Waters Corp Waters HPLC Column, Preparative, XTerra Shield RP8, 10 μm, 30 x 100 mm
Catalog No. 50781046 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XTerra HPLC Column is a preparative column featuring first-generation hybrid particle technology. It combines the high efficiency separations typically associated with silica-based materials with the expanded pH range of a polymer particle, offering a rugged material with high mechanical strength for various chromatographic applications.
  • Methylene bridged particles.
  • Combines high efficiency separations of silica with expanded pH range of a polymer particle.
  • Rugged material with high mechanical strength.
  • High efficiency.
  • Excellent peak shape for bases.
  • Wider pH range than silica-based particles (pH 1–12).
  • Easy scale-up from analytical to preparative chromatography.
  • Ability to operate at high pH provides excellent peak shapes for bases and increased mass loading (up to 60x) compared to conventional silica-based materials.
Catalog No. 50-781-046 Supplier Waters Corp Supplier No. 186002270
Add to Cart
Add to Cart
Request a Quote
The XTerra HPLC Column is a preparative column featuring first-generation hybrid particle technology. It combines the high efficiency separations typically associated with silica-based materials with the expanded pH range of a polymer particle, offering a rugged material with high mechanical strength for various chromatographic applications.
  • Methylene bridged particles.
  • Combines high efficiency separations of silica with expanded pH range of a polymer particle.
  • Rugged material with high mechanical strength.
  • High efficiency.
  • Excellent peak shape for bases.
  • Wider pH range than silica-based particles (pH 1–12).
  • Easy scale-up from analytical to preparative chromatography.
  • Ability to operate at high pH provides excellent peak shapes for bases and increased mass loading (up to 60x) compared to conventional silica-based materials.