Learn More
Waters Corp Prep Column, Shield RP8, 10 μm, 30 mm x 250 mm
Catalog No. 50781050 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XTerra Shield RP8 OBD Prep Column, 10 μm, 30 mm x 250 mm, combines the efficiency of silica with the stability of polymers for high-efficiency separations over an expanded pH range. It provides excellent peak shapes for bases, suitable for various chromatographic applications.
  • First-generation hybrid particle for silica efficiency and polymer stability.
  • Achieves high-efficiency separations with excellent peak shape for bases.
  • Provides expanded pH range (2–12) for rugged material.
  • Homogenous coverage and reduced silanols for superior peak shape.
  • Facilitates easy scale-up from analytical to preparative chromatography.
  • Supports high pH operation, increasing mass loading up to 60x.
  • Incorporates an embedded polar group for optimal peak shape and unique selectivity.
  • Exhibits excellent water wettability in 100% aqueous mobile phases.
Catalog No. 50-781-050 Supplier Waters Corp Supplier No. 186002274
Add to Cart
Add to Cart
Request a Quote
This XTerra Shield RP8 OBD Prep Column, 10 μm, 30 mm x 250 mm, combines the efficiency of silica with the stability of polymers for high-efficiency separations over an expanded pH range. It provides excellent peak shapes for bases, suitable for various chromatographic applications.
  • First-generation hybrid particle for silica efficiency and polymer stability.
  • Achieves high-efficiency separations with excellent peak shape for bases.
  • Provides expanded pH range (2–12) for rugged material.
  • Homogenous coverage and reduced silanols for superior peak shape.
  • Facilitates easy scale-up from analytical to preparative chromatography.
  • Supports high pH operation, increasing mass loading up to 60x.
  • Incorporates an embedded polar group for optimal peak shape and unique selectivity.
  • Exhibits excellent water wettability in 100% aqueous mobile phases.