Learn More
Waters Corp XTerra Shield RP8 Prep Column, 125Å, 10 µm, 7.8 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186001163
Encompass_Preferred
XTerra Preparative RP8 columns are hybrid-based, reversed-phase C8 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.
Catalog No. 50-780-595