Learn More
Waters Corp Waters XTerra Shield RP8 OBD Prep Column, 10 μm, 50 mm X 50 mm, 125 Å
Catalog No. 50781058 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Preparative RP8 columns are hybrid-based, reversed-phase C8 columns that incorporate patented embedded polar group technology. These columns are designed to improve peak shape performance for highly basic compounds and offer superior pH stability compared to traditional silica-based columns.
  • Hybrid-based, reversed-phase C8 columns
  • Incorporates patented embedded polar group technology
  • Designed to improve peak shape performance for highly basic compounds
  • Offers superior pH stability compared to silica-based columns
  • Spherical particle shape
  • Low silanol activity
  • Endcapped
  • Improved peak shape performance for highly basic compounds
  • Superior pH stability
Catalog No. 50-781-058 Supplier Waters Corp Supplier No. 186002282
Add to Cart
Add to Cart
Request a Quote
XTerra Preparative RP8 columns are hybrid-based, reversed-phase C8 columns that incorporate patented embedded polar group technology. These columns are designed to improve peak shape performance for highly basic compounds and offer superior pH stability compared to traditional silica-based columns.
  • Hybrid-based, reversed-phase C8 columns
  • Incorporates patented embedded polar group technology
  • Designed to improve peak shape performance for highly basic compounds
  • Offers superior pH stability compared to silica-based columns
  • Spherical particle shape
  • Low silanol activity
  • Endcapped
  • Improved peak shape performance for highly basic compounds
  • Superior pH stability