Learn More
Waters Corp Waters XTerra Shield RP8 Prep Column, 5 μm, 7.8 mm x 50 mm
Catalog No. 50780587 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Preparative RP8 columns are hybrid-based, reversed-phase C8 columns that were among the first commercially available hybrid particle stationary phases. They combine the high efficiency separations of a silica particle with the expanded pH range of a polymer particle, offering superior pH stability compared to silica-based columns.
  • Features hybrid particle design for high efficiency and stability.
  • Offers superior pH stability and a broad pH range (2 to 12).
  • Incorporates embedded polar group technology for improved peak shapes of basic compounds.
  • Designed for high efficiency separations.
  • Provides easy scale-up from analytical to preparative chromatography.
  • Allows for increased mass loading at high pH compared to conventional silica-based materials.
Catalog No. 50-780-587 Supplier Waters Corp Supplier No. 186001155
XTerra Preparative RP8 columns are hybrid-based, reversed-phase C8 columns that were among the first commercially available hybrid particle stationary phases. They combine the high efficiency separations of a silica particle with the expanded pH range of a polymer particle, offering superior pH stability compared to silica-based columns.
  • Features hybrid particle design for high efficiency and stability.
  • Offers superior pH stability and a broad pH range (2 to 12).
  • Incorporates embedded polar group technology for improved peak shapes of basic compounds.
  • Designed for high efficiency separations.
  • Provides easy scale-up from analytical to preparative chromatography.
  • Allows for increased mass loading at high pH compared to conventional silica-based materials.