Learn More
Waters Corp Waters HPLC Column, XTerra Shield RP8, 5 μm, 30 mm x 50 mm
Catalog No. 50780832 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra HPLC Columns are hybrid particle stationary phases, offering high efficiency, excellent peak shape, and an expanded pH range. This preparative column features Shield technology.
  • Hybrid particle (silica co-polymer) with high mechanical strength, efficiency, and excellent peak shape for bases
  • Wider pH range compared to silica-based particles, 2–12
  • Incorporates an embedded polar group for optimal peak shape and unique selectivity
  • Exhibits excellent water wettability, even in 100% aqueous mobile phases
  • Features Shield technology
  • Facilitates easy scale-up from analytical to preparative chromatography and enables operation at high pH for increased mass loading (up to 60x)
Catalog No. 50-780-832 Supplier Waters Corp Supplier No. 186001941
Add to Cart
Add to Cart
XTerra HPLC Columns are hybrid particle stationary phases, offering high efficiency, excellent peak shape, and an expanded pH range. This preparative column features Shield technology.
  • Hybrid particle (silica co-polymer) with high mechanical strength, efficiency, and excellent peak shape for bases
  • Wider pH range compared to silica-based particles, 2–12
  • Incorporates an embedded polar group for optimal peak shape and unique selectivity
  • Exhibits excellent water wettability, even in 100% aqueous mobile phases
  • Features Shield technology
  • Facilitates easy scale-up from analytical to preparative chromatography and enables operation at high pH for increased mass loading (up to 60x)