Learn More
Waters Corp HPLC Column, Shield RP8, 5 μm, 19 x 100 mm
Supplier:  Waters Corp 186001937
Encompass_Preferred
XTerra™ HPLC Columns are hybrid particle (silica co-polymer) stationary phases, combining the efficiency of silica with the stability of polymers. They are designed for high-efficiency separations, offering an expanded pH range and high mechanical strength. The XTerra RP phases, such as Shield RP8, incorporate an embedded polar group for optimal peak shape and unique selectivity, ensuring excellent water wettability in 100% aqueous mobile phases.
  • Hybrid particle (silica co-polymer) stationary phase
  • High efficiency separations
  • Expanded pH range
  • High mechanical strength
  • Excellent peak shape for bases
  • Wider pH range than silica-based particles
  • Embedded polar group for optimal peak shape and unique selectivity
  • Excellent water wettability in 100% aqueous mobile phases
Catalog No. 50-780-828