Learn More
Waters Corp Waters XTerra Shield RP18 Column, 125Å, 3.5 μm, 2.1 mm x 150 mm
Catalog No. 50780229 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These hybrid-based, reversed-phase C18 columns are engineered for the precise separation of diverse polar and non-polar compounds. They incorporate patented embedded polar group technology for improved peak shape performance for highly basic compounds and offer superior pH stability. With 125Å porous silica and a 3.5 μm particle size, they achieve remarkable separation efficiency.
  • Provides a wider pH range and superior mechanical strength compared to silica-based columns.
  • Enhances peak capacity and supports high flow rates with a 2.1 mm inner diameter and 150 mm length.
  • Includes a guard column for protection against contamination, reduced dead volume, and extended column lifetime.
  • Exhibits excellent water wettability, even in 100% aqueous mobile phases.
  • Compatible with various HPLC systems and detectors including UV, fluorescence, electrochemical, and RI.
Catalog No. 50-780-229 Supplier Waters Corp Supplier No. 186000410
These hybrid-based, reversed-phase C18 columns are engineered for the precise separation of diverse polar and non-polar compounds. They incorporate patented embedded polar group technology for improved peak shape performance for highly basic compounds and offer superior pH stability. With 125Å porous silica and a 3.5 μm particle size, they achieve remarkable separation efficiency.
  • Provides a wider pH range and superior mechanical strength compared to silica-based columns.
  • Enhances peak capacity and supports high flow rates with a 2.1 mm inner diameter and 150 mm length.
  • Includes a guard column for protection against contamination, reduced dead volume, and extended column lifetime.
  • Exhibits excellent water wettability, even in 100% aqueous mobile phases.
  • Compatible with various HPLC systems and detectors including UV, fluorescence, electrochemical, and RI.