Learn More
Waters Corp Waters HPLC Column, Reversed Phase, 3.5 μm, 125 Å, 3.9 mm x 100 mm
Catalog No. 50780247 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP18 columns are general-purpose, hybrid-based, reversed-phase C18 columns with embedded polar group technology, improving peak shape for highly basic compounds. They offer superior pH stability (2–12) compared to silica-based columns, utilizing Hybrid Particle Technology (HPT) for high mechanical strength, efficiency, and excellent peak shape.
  • Offers high mechanical strength and extremely spherical shape.
  • Achieves high efficiency and excellent peak shape for bases.
  • Manufactured in accordance with ISO 9001 guidelines.
  • Individually checked for performance, with a certificate of Batch Analysis and Column Efficiency.
Catalog No. 50-780-247 Supplier Waters Corp Supplier No. 186000428
XTerra RP18 columns are general-purpose, hybrid-based, reversed-phase C18 columns with embedded polar group technology, improving peak shape for highly basic compounds. They offer superior pH stability (2–12) compared to silica-based columns, utilizing Hybrid Particle Technology (HPT) for high mechanical strength, efficiency, and excellent peak shape.
  • Offers high mechanical strength and extremely spherical shape.
  • Achieves high efficiency and excellent peak shape for bases.
  • Manufactured in accordance with ISO 9001 guidelines.
  • Individually checked for performance, with a certificate of Batch Analysis and Column Efficiency.