Learn More
Waters Corp WATERS CORPORATION Column, 125Å, 3.5 μm, 4.6 mm x 250 mm
Catalog No. 50780701 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP18 columns are general-purpose, hybrid-based, reversed-phase C18 columns. They utilize embedded polar group technology for improved peak shape performance with highly basic compounds. Built on Hybrid Particle Technology, these columns offer superior pH stability from 2 to 12 compared to silica-based columns. This rugged organic/inorganic particle structure enables high-speed, high-temperature, and high-pH operation, with 33% fewer residual silanols for sharp, high-efficiency peaks.
  • General-purpose, hybrid-based, reversed-phase C18.
  • Improved peak shape for basic compounds.
  • Superior pH stability (2 to 12).
  • High-speed, high-temperature, high-pH operation.
  • Fewer residual silanols for high-efficiency peaks.
  • Manufactured with ultra-pure reagents.
  • Individually tested; CoA and CoE provided.
Catalog No. 50-780-701 Supplier Waters Corp Supplier No. 186001472
XTerra RP18 columns are general-purpose, hybrid-based, reversed-phase C18 columns. They utilize embedded polar group technology for improved peak shape performance with highly basic compounds. Built on Hybrid Particle Technology, these columns offer superior pH stability from 2 to 12 compared to silica-based columns. This rugged organic/inorganic particle structure enables high-speed, high-temperature, and high-pH operation, with 33% fewer residual silanols for sharp, high-efficiency peaks.
  • General-purpose, hybrid-based, reversed-phase C18.
  • Improved peak shape for basic compounds.
  • Superior pH stability (2 to 12).
  • High-speed, high-temperature, high-pH operation.
  • Fewer residual silanols for high-efficiency peaks.
  • Manufactured with ultra-pure reagents.
  • Individually tested; CoA and CoE provided.