Learn More
Waters Corp Waters XTerra Shield RP18 Column, 3.5 μm, 4.6 mm X 100 mm
Catalog No. 50780257 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP18 columns are general-purpose, hybrid-based, reversed-phase C18 columns designed to enhance peak shape performance for highly basic compounds. These columns offer superior pH stability compared to silica-based alternatives, overcoming the hydrolytic instability common with high pH applications by utilizing organic/inorganic Hybrid Particle Technology (HPT).
  • Enhances peak shape for highly basic compounds.
  • Provides superior pH stability (2 to 12).
  • Overcomes limitations of silica-based packing materials.
  • Features Hybrid Particle Technology (HPT) for mechanical strength and spherical shape.
  • Offers adjustable pore dimensions and surface area.
  • Manufactured under ISO 9001 conditions.
Catalog No. 50-780-257 Supplier Waters Corp Supplier No. 186000438
XTerra RP18 columns are general-purpose, hybrid-based, reversed-phase C18 columns designed to enhance peak shape performance for highly basic compounds. These columns offer superior pH stability compared to silica-based alternatives, overcoming the hydrolytic instability common with high pH applications by utilizing organic/inorganic Hybrid Particle Technology (HPT).
  • Enhances peak shape for highly basic compounds.
  • Provides superior pH stability (2 to 12).
  • Overcomes limitations of silica-based packing materials.
  • Features Hybrid Particle Technology (HPT) for mechanical strength and spherical shape.
  • Offers adjustable pore dimensions and surface area.
  • Manufactured under ISO 9001 conditions.