Learn More
Waters Corp Waters XTerra Shield RP18 Column, 3.5 μm, 4.6 mm, 50 mm
Supplier:  Waters Corp 186000434
Encompass_Preferred
XTerra RP18 columns are general purpose, hybrid-based, reversed-phase C18 columns that incorporate patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.
  • Incorporates patented embedded polar group technology to improve peak shape performance for highly basic compounds.
  • Provides superior pH stability compared to silica-based columns.
  • Versatile and capable of providing optimal shape performance for strongly basic chemicals while maintaining good separation efficiency and pH stability.
  • Wide operating pH range of 2 to 12.
  • Uses patented organic/inorganic Hybrid Particle Technology (HPT) to overcome limitations of silica-based reversed-phase packing materials, particularly hydrolytic instability at high pH.
Catalog No. 50-780-253