Learn More
Waters Corp XTerra HPLC Column, Shield RP18, 4.6 x 150 mm, 3.5 μm
Catalog No. 50818715 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
WATERS CORPORATION XTerra HPLC Columns are designed for high-performance liquid chromatography applications. The XTerra Shield RP18 column features a unique hybrid particle stationary phase, providing optimal performance and selectivity across a wide range of analytical conditions.
  • Embedded polar group for optimal peak shape and unique selectivity.
  • Excellent water wettability, even in 100% aqueous mobile phases.
  • Shield technology for the best peak shape for bases.
  • pH range of 2–12.
  • Ideally suited for UV, fluorescence, electrochemical, and RI detectors.
Catalog No. 50-818-715 Supplier Waters Corp Supplier No. 186000442
WATERS CORPORATION XTerra HPLC Columns are designed for high-performance liquid chromatography applications. The XTerra Shield RP18 column features a unique hybrid particle stationary phase, providing optimal performance and selectivity across a wide range of analytical conditions.
  • Embedded polar group for optimal peak shape and unique selectivity.
  • Excellent water wettability, even in 100% aqueous mobile phases.
  • Shield technology for the best peak shape for bases.
  • pH range of 2–12.
  • Ideally suited for UV, fluorescence, electrochemical, and RI detectors.