Learn More
Waters Corp XTerra Shield RP18 Column, 125Å, 5 µm, 2.1 mm X 100 mm, 1/pk
Supplier:  Waters Corp 186000452
Encompass_Preferred
XTerra RP18 columns are general purpose, hybrid-based, reversed-phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.
Catalog No. 50-780-269