Learn More
Waters Corp WATERS CORPORATION HPLC Column, Shield RP18, 5 μm, 3.0 × 250 mm
Catalog No. 50780293 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra HPLC Columns are first-generation hybrid particle columns designed to provide high-efficiency separations. They combine the efficiency of silica with the stability of polymers, offering an expanded pH range suitable for a broad range of applications.
  • Combines the efficiency of silica with the stability of polymers.
  • Offers high efficiency separations with an expanded pH range (1-12 or 2-12).
  • Possesses high mechanical strength.
  • Provides excellent peak shape for bases.
  • Features homogenous coverage and less silanols.
  • Facilitates easy scale-up to preparative chromatography.
  • Provides up to 60× increased mass loading.
Catalog No. 50-780-293 Supplier Waters Corp Supplier No. 186000476
XTerra HPLC Columns are first-generation hybrid particle columns designed to provide high-efficiency separations. They combine the efficiency of silica with the stability of polymers, offering an expanded pH range suitable for a broad range of applications.
  • Combines the efficiency of silica with the stability of polymers.
  • Offers high efficiency separations with an expanded pH range (1-12 or 2-12).
  • Possesses high mechanical strength.
  • Provides excellent peak shape for bases.
  • Features homogenous coverage and less silanols.
  • Facilitates easy scale-up to preparative chromatography.
  • Provides up to 60× increased mass loading.