Learn More
Waters Corp Waters XTerra Shield RP18 Column, 125Å, 5 μm, 3 mm x 50 mm
Supplier:  Waters Corp 186000464
Encompass_Preferred
XTerra RP18 columns are hybrid-based, reversed-phase C18 columns offering superior pH stability and embedded polar group technology for basic compounds. They feature a 125Å, 5 μm sorbent bed on an inert silica substrate for retention of polar and non-polar compounds. Ideal for pharmaceutical, bioanalytical, and environmental analyses.
  • 125Å, 5 μm sorbent bed for stable separation.
  • Retains polar and non-polar compounds.
  • Enhanced retention for basic compounds.
  • Robust design, easy to use, and extends LC system lifespan.
  • Cost-effective with high analytical precision.
  • Versatile for pharma, bio, and environmental applications.
Catalog No. 50-780-281