Learn More
Waters Corp WATERS CORPORATION Column, Shield RP18, 5 μm, 4.6 mm X 100 mm
Supplier:  Waters Corp 186000488
Encompass_Preferred
XTerra RP18 columns are general-purpose, hybrid-based, reversed-phase C18 columns that utilize Waters patented organic/inorganic Hybrid Particle Technology (HPT). They provide superior pH stability compared to silica-based columns and improve peak shape performance for highly basic compounds.
  • Improved peak shape performance for highly basic compounds due to patented embedded polar group technology.
  • Superior pH stability (2 to 12) compared to silica-based columns.
  • Overcomes hydrolytic instability of silica-based materials at high pH.
  • High mechanical strength and excellent peak shape for bases.
  • Exhibits excellent water wettability, even in 100% aqueous mobile phases.
  • Manufactured under ISO 9001 conditions, individually tested for performance.
Catalog No. 50-780-305