Learn More
Waters Corp Waters XTerra Shield RP18 Column, 5 μm, 4.6 mm x 50 mm
Catalog No. 50780301 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP18 columns are hybrid-based, reversed-phase C18 columns featuring embedded polar group technology for improved peak shape performance with highly basic compounds. They provide superior pH stability compared to silica-based columns and are manufactured to ISO 9001 standards.
  • Embedded polar group technology improves peak shape performance for highly basic compounds.
  • Superior pH stability offers a broad pH range of 2 to 12 pH.
  • Hybrid particle synthesis uses a high purity methyl hybrid material made from TEOS and MTEOS.
  • Achieves highly symmetrical and efficient peaks by reducing silanol interactions with basic analytes.
  • Robust separations are possible over a wide pH range and with various solvents.
  • Built to ISO 9001 quality standards.
Catalog No. 50-780-301 Supplier Waters Corp Supplier No. 186000484
Add to Cart
Add to Cart
XTerra RP18 columns are hybrid-based, reversed-phase C18 columns featuring embedded polar group technology for improved peak shape performance with highly basic compounds. They provide superior pH stability compared to silica-based columns and are manufactured to ISO 9001 standards.
  • Embedded polar group technology improves peak shape performance for highly basic compounds.
  • Superior pH stability offers a broad pH range of 2 to 12 pH.
  • Hybrid particle synthesis uses a high purity methyl hybrid material made from TEOS and MTEOS.
  • Achieves highly symmetrical and efficient peaks by reducing silanol interactions with basic analytes.
  • Robust separations are possible over a wide pH range and with various solvents.
  • Built to ISO 9001 quality standards.