Learn More
Waters Corp Waters Column, C18, 5 μm, 4.6 mm x 150 mm
Catalog No. 50819081 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP18 columns are general purpose, hybrid-based, reversed-phase C18 columns that utilize patented embedded polar group technology for enhanced peak shape performance of basic compounds. They offer superior pH stability (2 to 12 pH) compared to silica-based columns and combine Hybrid Particle Technology with Shield Technology for optimal peak shape and excellent water wettability.
  • Enhances peak shape for basic compounds with patented embedded polar group technology.
  • Provides superior pH stability, operating from 2 to 12 pH.
  • General purpose, hybrid-based, reversed-phase C18 for wide applicability.
  • Facilitates fast method development with unique selectivity.
  • Exhibits excellent water wettability, even in 100% aqueous mobile phases.
  • Rugged hybrid particle technology allows for high-speed, high-temperature, and high-pH operation.
Catalog No. 50-819-081 Supplier Waters Corp Supplier No. 186000492
XTerra RP18 columns are general purpose, hybrid-based, reversed-phase C18 columns that utilize patented embedded polar group technology for enhanced peak shape performance of basic compounds. They offer superior pH stability (2 to 12 pH) compared to silica-based columns and combine Hybrid Particle Technology with Shield Technology for optimal peak shape and excellent water wettability.
  • Enhances peak shape for basic compounds with patented embedded polar group technology.
  • Provides superior pH stability, operating from 2 to 12 pH.
  • General purpose, hybrid-based, reversed-phase C18 for wide applicability.
  • Facilitates fast method development with unique selectivity.
  • Exhibits excellent water wettability, even in 100% aqueous mobile phases.
  • Rugged hybrid particle technology allows for high-speed, high-temperature, and high-pH operation.