Learn More
Waters Corp HPLC Column, XTerra Shield RP18, 5 μm, 4.6 mm X 250 mm, 2–12 pH Range
Supplier:  Waters Corp 186000496
Encompass_Preferred
XTerra RP18 columns are general purpose, hybrid-based, reversed-phase C18 columns that incorporate patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.
  • General purpose, hybrid-based, reversed-phase C18 column
  • Incorporates patented embedded polar group technology to improve peak shape performance for highly basic compounds
  • Superior pH stability compared to a silica-based column
Catalog No. 50-818-716