Learn More
Waters Corp XTerra Hybrid Shield RP18 Method Validation Kit, 125Å, 5 µm, 4.6 mm X 150 mm, 3/pk
Supplier:  Waters Corp 186000862
Encompass_Preferred
This kit consists of three XTerra RP 18 columns packed with three different batches of sorbent.
Catalog No. 50-780-509