Learn More
Waters Corp Waters Sentry Guard Cartridge, Shield RP18, 3.5 μm, 3.9 mm x 20 mm
Catalog No. 50780435 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sentry Guard Cartridge is designed for reversed-phase liquid chromatography, for use with compatible columns. It attaches directly to the analytical column's inlet, safeguarding against chemical or particle contaminants and improving analytical performance by removing impurities from the mobile phase stream.
  • Produced to stringent standards.
  • Evaluated for optimal function, selectivity, sensitivity, and speed.
  • Made from premium components, matching compatible RP18 columns.
  • Manufactured in cGMP, ISO 9001-certified facility.
  • Packing material chromatographically tested per batch.
  • Offers improved pH stability versus silica-based columns.
  • Operates across a 2 to 12 pH range.
  • Max pressure tolerance of 6,000 psi (415 Bar).
Catalog No. 50-780-435 Supplier Waters Corp Supplier No. 186000646
This Sentry Guard Cartridge is designed for reversed-phase liquid chromatography, for use with compatible columns. It attaches directly to the analytical column's inlet, safeguarding against chemical or particle contaminants and improving analytical performance by removing impurities from the mobile phase stream.
  • Produced to stringent standards.
  • Evaluated for optimal function, selectivity, sensitivity, and speed.
  • Made from premium components, matching compatible RP18 columns.
  • Manufactured in cGMP, ISO 9001-certified facility.
  • Packing material chromatographically tested per batch.
  • Offers improved pH stability versus silica-based columns.
  • Operates across a 2 to 12 pH range.
  • Max pressure tolerance of 6,000 psi (415 Bar).