Learn More
Waters Corp XTerra Shield RP18 Sentry Guard Cartridge, 125Å, 5 µm, 3 mm X 20 mm, 2/pk
Catalog No. 50780447 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP18 columns are general purpose, hybrid-based, reversed-phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.
Catalog No. 50-780-447 Supplier Waters Corp Supplier No. 186000658
XTerra RP18 columns are general purpose, hybrid-based, reversed-phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.