Learn More
Waters Corp Waters Intelligent Speed (IS) Column, Shield RP18, 3.5 μm, 125 Å, 4.6 mm X 20 mm
Catalog No. 50780794 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP18 columns are general-purpose, hybrid-based, reversed-phase C18 columns designed to enhance peak shape performance for highly basic compounds. They offer superior pH stability compared to silica-based columns, excellent water wettability, and are suited for various detectors including UV, fluorescence, electrochemical, and RI.
  • Hybrid-based C18 chemistry with Shield RP18 bonding technology.
  • Particle size of 3.5 μm and pore size of 125 Å.
  • pH range of 2 to 12 and temperature limits of 20°C to 60°C.
  • Operates at up to 6000 psi (415 Bar) for HPLC systems.
  • Embedded polar group provides optimal peak shape, unique selectivity, and high efficiency separations.
  • Wider pH range allows for greater selectivity, improved peak shape, and increased mass loading.
  • Facilitates easy scale-up from analytical to preparative chromatography.
Catalog No. 50-780-794 Supplier Waters Corp Supplier No. 186001893
XTerra RP18 columns are general-purpose, hybrid-based, reversed-phase C18 columns designed to enhance peak shape performance for highly basic compounds. They offer superior pH stability compared to silica-based columns, excellent water wettability, and are suited for various detectors including UV, fluorescence, electrochemical, and RI.
  • Hybrid-based C18 chemistry with Shield RP18 bonding technology.
  • Particle size of 3.5 μm and pore size of 125 Å.
  • pH range of 2 to 12 and temperature limits of 20°C to 60°C.
  • Operates at up to 6000 psi (415 Bar) for HPLC systems.
  • Embedded polar group provides optimal peak shape, unique selectivity, and high efficiency separations.
  • Wider pH range allows for greater selectivity, improved peak shape, and increased mass loading.
  • Facilitates easy scale-up from analytical to preparative chromatography.