Learn More
Waters Corp Waters Column, XTerra Shield RP18, 3.5 μm, 3.9 mm x 20 mm
Catalog No. 50780801 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP18 columns are general purpose, hybrid-based, reversed-phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first-generation columns provide superior pH stability compared to a silica-based column.
  • Hybrid particle technology provides high mechanical strength and efficiency, supporting 6000 psi (415 bar).
  • Expanded pH range (2 to 12) for greater selectivity and improved peak shape.
  • Embedded polar group ensures excellent water wettability in 100% aqueous mobile phases.
  • Delivers superior peak shape with homogenous coverage and fewer silanols.
  • Suitable for mass spectrometry, UV, fluorescence, electrochemical, and RI detection.
  • Supports easy scale-up from analytical to preparative chromatography.
  • Features spherical particle shape and 125 Å pore size.
Catalog No. 50-780-801 Supplier Waters Corp Supplier No. 186001902
Add to Cart
Add to Cart
XTerra RP18 columns are general purpose, hybrid-based, reversed-phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first-generation columns provide superior pH stability compared to a silica-based column.
  • Hybrid particle technology provides high mechanical strength and efficiency, supporting 6000 psi (415 bar).
  • Expanded pH range (2 to 12) for greater selectivity and improved peak shape.
  • Embedded polar group ensures excellent water wettability in 100% aqueous mobile phases.
  • Delivers superior peak shape with homogenous coverage and fewer silanols.
  • Suitable for mass spectrometry, UV, fluorescence, electrochemical, and RI detection.
  • Supports easy scale-up from analytical to preparative chromatography.
  • Features spherical particle shape and 125 Å pore size.