Learn More
Waters Corp XTerra Shield RP8 Column, 125Å, 3.5 µm, 2.1 mm X 150 mm, 1/pk
Catalog No. 50780230 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP8 columns are general purpose, hybrid-based, reversed-phase C8 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.
Catalog No. 50-780-230 Supplier Waters Corp Supplier No. 186000411
XTerra RP8 columns are general purpose, hybrid-based, reversed-phase C8 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.