Learn More
Waters Corp Waters Corporation XTerra Shield RP8 Column, 3.5 μm, 2.1 x 100 mm
Catalog No. 50780226 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This general-purpose, hybrid-based, reversed-phase C8 column employs embedded polar group technology, enhancing peak shape for highly basic compounds. It offers superior pH stability over silica-based columns and pioneered hybrid particle stationary phases, combining efficiency, mechanical strength, and broad pH range.
  • Embedded polar group technology improves peak shape for highly basic compounds.
  • Superior pH stability compared to silica-based columns.
  • Hybrid particle technology combines high efficiency with expanded pH range.
  • Wide pH range of 2–12 for versatile separations.
  • High mechanical strength for a robust and durable material.
  • Excellent water wettability, even in 100% aqueous mobile phases.
Catalog No. 50-780-226 Supplier Waters Corp Supplier No. 186000407
This general-purpose, hybrid-based, reversed-phase C8 column employs embedded polar group technology, enhancing peak shape for highly basic compounds. It offers superior pH stability over silica-based columns and pioneered hybrid particle stationary phases, combining efficiency, mechanical strength, and broad pH range.
  • Embedded polar group technology improves peak shape for highly basic compounds.
  • Superior pH stability compared to silica-based columns.
  • Hybrid particle technology combines high efficiency with expanded pH range.
  • Wide pH range of 2–12 for versatile separations.
  • High mechanical strength for a robust and durable material.
  • Excellent water wettability, even in 100% aqueous mobile phases.