Learn More
Waters Corp Waters XTerra Shield RP8 Column, 125Å, 3.5 μm, 4.6 mm x 100 mm
Supplier:  Waters Corp 186000439
Encompass_Preferred
XTerra Shield RP8 Columns are first-generation, hybrid-based, reversed-phase C8 columns. They combine the efficiency of silica particles with the enhanced pH stability of polymer particles. Designed with patented embedded polar group technology, these columns improve peak shape performance for highly basic compounds and offer superior pH stability compared to silica-based columns, featuring a widened usable pH range of 2–12.
  • Hybrid particle technology combines silica efficiency with polymer stability.
  • High mechanical strength and efficiency.
  • Excellent peak shape for basic compounds.
  • Wide pH range of 2–12.
  • Reduced silanols compared to other materials.
  • Endcapped for consistent performance.
  • Spherical particle shape.
  • Suitable for HPLC systems.
  • USP classification L7.
  • Carbon load of 14%.
Catalog No. 50-780-258