Learn More
Waters Corp Waters Column, XTerra Shield RP8, 3.5 μm, 4.6 mm x 150 mm, 125 Å
Catalog No. 50818718 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP8 columns are hybrid-based, reversed-phase C8 columns designed for superior pH stability and improved peak shape performance for basic compounds. They utilize patented Hybrid Particle Technology for high-speed separations at high temperatures and pH, offering exceptionally sharp and efficient peaks due to fewer residual silanols.
  • Incorporates patented embedded polar group technology for improved peak shape with basic compounds.
  • Provides superior pH stability compared to silica-based columns.
  • Uses Hybrid Particle Technology for high-speed separations at high temperatures and high pH.
  • Features 33% fewer residual silanols for exceptionally sharp and efficient peaks.
  • Manufactured with ultrapure reagents; each column is individually tested with a quality control report.
  • Lifetime can be extended with a VanGuard Cartridge.
Catalog No. 50-818-718 Supplier Waters Corp Supplier No. 186000443
XTerra RP8 columns are hybrid-based, reversed-phase C8 columns designed for superior pH stability and improved peak shape performance for basic compounds. They utilize patented Hybrid Particle Technology for high-speed separations at high temperatures and pH, offering exceptionally sharp and efficient peaks due to fewer residual silanols.
  • Incorporates patented embedded polar group technology for improved peak shape with basic compounds.
  • Provides superior pH stability compared to silica-based columns.
  • Uses Hybrid Particle Technology for high-speed separations at high temperatures and high pH.
  • Features 33% fewer residual silanols for exceptionally sharp and efficient peaks.
  • Manufactured with ultrapure reagents; each column is individually tested with a quality control report.
  • Lifetime can be extended with a VanGuard Cartridge.