Learn More
Waters Corp XTerra Shield RP8 Column, C8, 5 μm, 2.1 mm × 100 mm
Catalog No. 50780270 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP8 columns are general-purpose, hybrid-based, reversed-phase C8 columns with patented embedded polar group technology, improving peak shape for highly basic compounds. They offer superior pH stability over silica-based columns, providing mechanical strength, spherical shape, and high purity.
  • Improves peak shape for highly basic compounds.
  • Superior pH stability.
  • Excellent separation efficiency.
  • Optimal shape performance for basic chemicals.
  • Large pH range (2 to 12 pH).
  • Utilizes proprietary hybrid particle technology (HPT) for hydrolytic stability at high pH.
  • Constructed from high-purity methyl hybrid material.
  • Produced to ISO 9001 guidelines.
  • Individually evaluated for performance.
  • Certificates of batch analysis and column efficiency provided.
Catalog No. 50-780-270 Supplier Waters Corp Supplier No. 186000453
XTerra RP8 columns are general-purpose, hybrid-based, reversed-phase C8 columns with patented embedded polar group technology, improving peak shape for highly basic compounds. They offer superior pH stability over silica-based columns, providing mechanical strength, spherical shape, and high purity.
  • Improves peak shape for highly basic compounds.
  • Superior pH stability.
  • Excellent separation efficiency.
  • Optimal shape performance for basic chemicals.
  • Large pH range (2 to 12 pH).
  • Utilizes proprietary hybrid particle technology (HPT) for hydrolytic stability at high pH.
  • Constructed from high-purity methyl hybrid material.
  • Produced to ISO 9001 guidelines.
  • Individually evaluated for performance.
  • Certificates of batch analysis and column efficiency provided.