Learn More
Waters Corp Waters Column, Shield RP8, 5 μm, 2.1 mm ID, 250 mm Length
Catalog No. 50780278 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra™ HPLC Columns are hybrid particle stationary phases combining silica efficiency with polymer stability. These reversed-phase C8 columns feature methylene-bridged silica co-polymer particles and embedded polar group technology, improving peak shape for basic compounds and offering superior pH stability.
  • General purpose, hybrid-based, reversed-phase C8 column chemistry.
  • Embedded polar group technology for improved peak shape of basic compounds.
  • Superior pH stability with a range of 2–12.
  • High mechanical strength, efficiency, and spherical particle shape.
  • Optimized dimensions: 5 μm particle size, 125 Å pore size, 2.1 mm ID, 250 mm length.
  • Maximum pressure up to 6000 psi (415 Bar).
  • Endcapped with low silanol activity.
  • Suitable for HPLC systems and easy scale-up.
Catalog No. 50-780-278 Supplier Waters Corp Supplier No. 186000461
Add to Cart
Add to Cart
XTerra™ HPLC Columns are hybrid particle stationary phases combining silica efficiency with polymer stability. These reversed-phase C8 columns feature methylene-bridged silica co-polymer particles and embedded polar group technology, improving peak shape for basic compounds and offering superior pH stability.
  • General purpose, hybrid-based, reversed-phase C8 column chemistry.
  • Embedded polar group technology for improved peak shape of basic compounds.
  • Superior pH stability with a range of 2–12.
  • High mechanical strength, efficiency, and spherical particle shape.
  • Optimized dimensions: 5 μm particle size, 125 Å pore size, 2.1 mm ID, 250 mm length.
  • Maximum pressure up to 6000 psi (415 Bar).
  • Endcapped with low silanol activity.
  • Suitable for HPLC systems and easy scale-up.