Learn More
Waters Corp Waters XTerra Shield RP8 Column, 5 μm, 2.1 mm x 50 mm
Catalog No. 50780266 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP8 columns are general purpose, hybrid-based, reversed-phase C8 columns that incorporate embedded polar group technology to improve peak shape performance for highly basic compounds. These columns offer superior pH stability compared to silica-based columns.
  • Hybrid particle technology provides high efficiency and extended pH range.
  • Improved peak shape performance for basic compounds due to embedded polar group.
  • Superior pH stability compared to silica-based columns.
  • High mechanical strength and rugged material for robustness.
  • Offers unique selectivity and excellent water wettability in aqueous mobile phases.
  • Facilitates easy scale-up from analytical to preparative chromatography.
Catalog No. 50-780-266 Supplier Waters Corp Supplier No. 186000449
XTerra RP8 columns are general purpose, hybrid-based, reversed-phase C8 columns that incorporate embedded polar group technology to improve peak shape performance for highly basic compounds. These columns offer superior pH stability compared to silica-based columns.
  • Hybrid particle technology provides high efficiency and extended pH range.
  • Improved peak shape performance for basic compounds due to embedded polar group.
  • Superior pH stability compared to silica-based columns.
  • High mechanical strength and rugged material for robustness.
  • Offers unique selectivity and excellent water wettability in aqueous mobile phases.
  • Facilitates easy scale-up from analytical to preparative chromatography.