Learn More
Waters Corp Waters XTerra Shield RP8 Column, 4.6 x 30 mm, 5 μm
Supplier:  Waters Corp 186001911
Encompass_Preferred
XTerra RP Columns, including the RP8, incorporate an embedded polar group to deliver the best possible peak shape and unique selectivity. They exhibit excellent water wettability, even in 100% aqueous mobile phases.
  • Best peak shape for bases
  • Shield technology
  • pH range 2–12
  • Ideally suited for UV, fluorescence, electrochemical, and RI detectors
Catalog No. 50-780-807