Learn More
Waters Corp Waters Column, XTerra Shield RP8, 125Å, 5 μm, 4.6 mm x 50 mm
Supplier: Waters Corp 186000485
XTerra RP8 columns are general-purpose, hybrid-based, reversed-phase C8 columns that utilize patented embedded polar group technology for improved peak shape performance with highly basic compounds. They offer superior pH stability over silica-based columns, employing proprietary organic/inorganic hybrid particle technology (HPT) to mitigate hydrolytic instability at high pH. Constructed from high-purity methyl hybrid material, these ISO 9001-compliant columns are independently evaluated, with batch analysis and efficiency certificates included.
- Mechanical strength
- Highly spherical shape
- Purity
- Pore size
- Surface area
- Capacity to customize particle size
- Retains excellent separation efficiency
- Provides optimal shape performance for extremely basic chemicals
- Increased pH stability
- Works with a wide pH range