Learn More
Waters Corp XTerra Shield RP18, 125 Å, 5 μm, 4.6 × 100 mm
Catalog No. 502392232 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP18 columns are general-purpose, hybrid-based, reversed-phase C18 columns that incorporate embedded polar group technology to improve peak shape performance for highly basic compounds. These columns provide superior pH stability compared to a silica-based column. Equipped with an eConnect column tag, these NFC-enabled columns automatically identify, track, and digitally trace column use history. eConnect columns can be used on any HPLC, but their advantages are unlocked when used with the Alliance iS System running Empower software.
  • Automatically identify, track, and digitally trace column use history.
  • Can be used on any HPLC.
  • Advantages are unlocked when used with the Alliance iS System running Empower software.
  • Superior pH stability compared to a silica-based column.
  • Improved peak shape performance for highly basic compounds.
Catalog No. 50-239-2232 Supplier Waters Corp Supplier No. 186000488RF
XTerra RP18 columns are general-purpose, hybrid-based, reversed-phase C18 columns that incorporate embedded polar group technology to improve peak shape performance for highly basic compounds. These columns provide superior pH stability compared to a silica-based column. Equipped with an eConnect column tag, these NFC-enabled columns automatically identify, track, and digitally trace column use history. eConnect columns can be used on any HPLC, but their advantages are unlocked when used with the Alliance iS System running Empower software.
  • Automatically identify, track, and digitally trace column use history.
  • Can be used on any HPLC.
  • Advantages are unlocked when used with the Alliance iS System running Empower software.
  • Superior pH stability compared to a silica-based column.
  • Improved peak shape performance for highly basic compounds.