Learn More
Waters Corp HPLC Column, 3.5 μm Particle, 125 Å Pore, 4.6 mm x 50 mm
Catalog No. 502391352 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra RP18 columns are general-purpose, hybrid-based, reversed-phase C18 columns. They incorporate patented embedded polar group technology to enhance peak shape performance for highly basic compounds and offer superior pH stability compared to silica-based columns.
  • Features an electronic tag with unique, product-specific identifying information.
  • NFC-enabled for automatic identification, tracking, and digital tracing of column use history.
  • Tag data automatically imports and is searchable via the HPLC system's software.
  • Pre-equipped tag simplifies setup and reduces manual data entry.
  • Permanent, tamper-proof design enhances compliance and auto-detectability.
  • Optimized tag placement prevents interference in temperature-sensitive methods.
  • NFC capability provides convenient access to product-support information.
Catalog No. 50-239-1352 Supplier Waters Corp Supplier No. 186000434RF
XTerra RP18 columns are general-purpose, hybrid-based, reversed-phase C18 columns. They incorporate patented embedded polar group technology to enhance peak shape performance for highly basic compounds and offer superior pH stability compared to silica-based columns.
  • Features an electronic tag with unique, product-specific identifying information.
  • NFC-enabled for automatic identification, tracking, and digital tracing of column use history.
  • Tag data automatically imports and is searchable via the HPLC system's software.
  • Pre-equipped tag simplifies setup and reduces manual data entry.
  • Permanent, tamper-proof design enhances compliance and auto-detectability.
  • Optimized tag placement prevents interference in temperature-sensitive methods.
  • NFC capability provides convenient access to product-support information.