Learn More
Waters Corp HPLC Column, XTerra Shield RP18, 5 μm, 10 x 100 mm
Catalog No. 502391439 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These preparative RP18 columns are hybrid-based, reversed-phase C18 columns designed to improve peak shape performance for highly basic compounds and offer superior pH stability compared to silica-based columns.
  • Hybrid-based, reversed-phase C18 column chemistry.
  • Embedded polar group technology improves peak shape of highly basic compounds.
  • Superior pH stability (range 2 to 12) versus silica-based columns.
  • High efficiency separations with spherical 5 μm particles and 125 Å pore size.
  • Rugged design, high mechanical strength, proprietary end-capping; stable 20°C to 60°C.
  • Excellent water wettability; compatible with UV, fluorescence, electrochemical, RI detectors.
  • Easy scale-up; increased mass loading (up to 60x) and enhanced selectivity at high pH.
Catalog No. 50-239-1439 Supplier Waters Corp Supplier No. 186008128
These preparative RP18 columns are hybrid-based, reversed-phase C18 columns designed to improve peak shape performance for highly basic compounds and offer superior pH stability compared to silica-based columns.
  • Hybrid-based, reversed-phase C18 column chemistry.
  • Embedded polar group technology improves peak shape of highly basic compounds.
  • Superior pH stability (range 2 to 12) versus silica-based columns.
  • High efficiency separations with spherical 5 μm particles and 125 Å pore size.
  • Rugged design, high mechanical strength, proprietary end-capping; stable 20°C to 60°C.
  • Excellent water wettability; compatible with UV, fluorescence, electrochemical, RI detectors.
  • Easy scale-up; increased mass loading (up to 60x) and enhanced selectivity at high pH.