Learn More
Waters Corp Waters XTerra Shield RP18 Column, 10 x 50 mm, 5 μm, 125 Å
Catalog No. 502038338 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This preparative HPLC column combines high-efficiency silica particle separations with the expanded pH range of a polymer particle. It provides rugged materials, high mechanical strength, and excellent peak shape for bases across a wider pH range than traditional silica-based particles.
  • Incorporates an embedded polar group for optimal peak shape and unique selectivity.
  • Excellent water wettability, even in 100% aqueous mobile phases.
  • Bonded and endcapped particles with homogenous coverage, fewer silanols, superior peak shape, and a pH range of 1–12.
  • Suited for UV, fluorescence, electrochemical, and RI detectors.
  • Easy scale-up from analytical to preparative chromatography.
  • High pH operation yields excellent peak shapes for bases and increased mass loading (up to 60x).
Catalog No. 50-203-8338 Supplier Waters Corp Supplier No. 186008105
This preparative HPLC column combines high-efficiency silica particle separations with the expanded pH range of a polymer particle. It provides rugged materials, high mechanical strength, and excellent peak shape for bases across a wider pH range than traditional silica-based particles.
  • Incorporates an embedded polar group for optimal peak shape and unique selectivity.
  • Excellent water wettability, even in 100% aqueous mobile phases.
  • Bonded and endcapped particles with homogenous coverage, fewer silanols, superior peak shape, and a pH range of 1–12.
  • Suited for UV, fluorescence, electrochemical, and RI detectors.
  • Easy scale-up from analytical to preparative chromatography.
  • High pH operation yields excellent peak shapes for bases and increased mass loading (up to 60x).