Learn More
Waters Corp XTerra Prep Guard Cartridge, Shield RP18, 30 mm x 10 mm
Catalog No. 502392424 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 30x10mm Prep Guard Cartridge protects HPLC and SFC 30 mm ID Prep OBD™ columns, extending column lifetimes and reducing purification costs when placed ahead of the prep column. It integrates into existing analytical workflows, offering enhanced protection from contaminants. It features the same diameter as associated preparative columns for a seamless experience and is packed with the same sorbent.
  • Protects HPLC and SFC 30 mm ID Prep OBD™ columns
  • Extends column lifetimes and reduces purification costs
  • Integrates into existing analytical workflows, enhancing contaminant protection
  • Matches diameter of associated preparative columns and uses same sorbent
  • Acts as a barrier against mobile phase particulate contaminants
  • Provides accurate data and reduces separation reproducibility needs
  • Withstands high backpressures up to 6,000 PSI for robust performance
Catalog No. 50-239-2424 Supplier Waters Corp Supplier No. 186006906
Add to Cart
Add to Cart
The 30x10mm Prep Guard Cartridge protects HPLC and SFC 30 mm ID Prep OBD™ columns, extending column lifetimes and reducing purification costs when placed ahead of the prep column. It integrates into existing analytical workflows, offering enhanced protection from contaminants. It features the same diameter as associated preparative columns for a seamless experience and is packed with the same sorbent.
  • Protects HPLC and SFC 30 mm ID Prep OBD™ columns
  • Extends column lifetimes and reduces purification costs
  • Integrates into existing analytical workflows, enhancing contaminant protection
  • Matches diameter of associated preparative columns and uses same sorbent
  • Acts as a barrier against mobile phase particulate contaminants
  • Provides accurate data and reduces separation reproducibility needs
  • Withstands high backpressures up to 6,000 PSI for robust performance